Anti-GAD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20746-1-AP-150UL
10087-302EA 402 USD
10087-302
Anti-GAD2 Rabbit Polyclonal Antibody
Antibodies
GAD2, also named as GAD65, belongs to the group II decarboxylase family. GAD2 catalyzes the production of GABA. It is responsible for the synthesis of the essential neurotransmitter gamma-aminobutyric acid (GABA) from L-glutamic acid. GAD2 is expressed in nervous and endocrine systems and are thought to be involved in synaptic transmission and insulin secretion. Autoantibodies against GAD2 may serve as markers for type I diabetes. Many individuals suffering from an adult onset disorder known as Stiff Person Syndrome (SPS) also express autoantibodies to GAD2. The antibody is specific to GAD2.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IF: HeLa Cells, 1:10-1:100

Type: Primary
Antigen: GAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR