Anti-Brachyury-T Rabbit Polyclonal Antibody

Supplier: Proteintech
20741-1-AP-150UL
10083-662EA 402 USD
10083-662
Anti-Brachyury-T Rabbit Polyclonal Antibody
Antibodies
Brachyury T is involved in the transcriptional regulation of genes required for mesoderm formation and differentiation. It binds to a palindromic site (called T site) and activates gene transcription when bound to such a site. Genetic variations in T are associated with susceptibility to neural tube defects (NTD). The antibody is specific to Brachyury T.

Western Blot: Mouse Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: Brachyury-T
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR