Anti-SYNM Rabbit Polyclonal Antibody

Supplier: Proteintech
20735-1-AP-150UL
10095-514EA 402 USD
10095-514
Anti-SYNM Rabbit Polyclonal Antibody
Antibodies
SYNM, also named as DMN, KIAA0353 and SYN, belongs to the intermediate filament family. SYNM is a Type-VI intermediate filament (IF) which plays an important cytoskeletal role within the muscle cell cytoskeleton. It forms heteropolymeric IFs with desmin and/or vimentin, and via its interaction with cytoskeletal proteins alpha-dystrobrevin, dystrophin, talin-1, utrophin and vinculin, is able to link these heteropolymeric IFs to adherens-type junctions, such as to the costameres, neuromuscular junctions, and myotendinous junctions within striated muscle cells. The antibody can’t recognize isofor2 and 3 of SYNM.

Western Blot:Y79 Cells, 1:500-1:5000; IHC: Human Skeletal Muscle Tissue, 1:20-1:200; IF: HeLa Cells, 1:10-1:100

Type: Primary
Antigen: SYNM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR