Anti-GNRHR2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20728-1-AP-150UL
10087-260EA 402 USD
10087-260
Anti-GNRHR2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200; IF: HEK-293 Cells, 1:10-1:100

Type: Primary
Antigen: GNRHR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR