Anti-CLDN22 Rabbit Polyclonal Antibody

Supplier: Proteintech
20724-1-AP-150UL
10084-830EA 402 USD
10084-830
Anti-CLDN22 Rabbit Polyclonal Antibody
Antibodies
CLDN22 belongs to the claudin family. It plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity. The antibody is specific to CLD22.

Western Blot:A431 Cells, 1:200-1:2000

Type: Primary
Antigen: CLDN22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR