Anti-TMC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20718-1-AP-150UL
10096-058EA 402 USD
10096-058
Anti-TMC1 Rabbit Polyclonal Antibody
Antibodies
TMC1 belongs to the TMC family. It is required for the normal function of cochlear hair cells. Defects in TMC1 are the cause of deafness autosomal dominant type 36 (DFNA36). Defects in TMC1 are the cause of deafness autosomal recessive type 7 (DFNB7). The antibody is specific to TMC1.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: TMC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR