Anti-ESPN Rabbit Polyclonal Antibody

Supplier: Proteintech
20717-1-AP-150UL
10086-654EA 402 USD
10086-654
Anti-ESPN Rabbit Polyclonal Antibody
Antibodies
ESPN, also named as DFNB36, is a multifunctional actin-bundling protein. ESPN plays a major role in regulating the organization, dimensions, dynamics and signaling capacities of the actin filament-rich, microvillus-type specializations that mediate sensory transduction in variouS mechanosensory and chemosensory cells. Defects in ESPN are the cause of deafness autosomal recessive type 36 (DFNB36). Defects in ESPN are the cause of deafness autosomal dominant without vestibular involvement (DFNAWVI). The antibody is specific to ESPN.

Western Blot: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: ESPN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR