Anti-MYH14 Rabbit Polyclonal Antibody

Supplier: Proteintech
20716-1-AP-150UL
10090-698EA 402 USD
10090-698
Anti-MYH14 Rabbit Polyclonal Antibody
Antibodies
MYH14, also named as KIAA2034 and NMHC II-C, is cellular myosin that appears to play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping. Defects in MYH14 are the cause of deafness autosomal dominant type 4 (DFNA4). The antibody is specific to MYH14.

Western Blot:COLO 320 Cells, 1:500-1:5000; IF: Hela, 1:10-1:100

Type: Primary
Antigen: MYH14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR