Anti-GHRHR Rabbit Polyclonal Antibody

Supplier: Proteintech
20715-1-AP-150UL
10087-536EA 402 USD
10087-536
Anti-GHRHR Rabbit Polyclonal Antibody
Antibodies
GHRHR, also named as GRFR, belongs to the G-protein coupled receptor 2 family. It is a receptor for GRF, coupled to G proteins which activate adenylyl cyclase. GHRHR stimulates somatotroph cell growth, growth hormone gene transcription and growth hormone secretion. The antibody is specific to GHRHR. The antibody is specific to GHRHR.

Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: GHRHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR