Anti-FLT4 Rabbit Polyclonal Antibody

Supplier: Proteintech
20712-1-AP-150UL
10096-958EA 402 USD
10096-958
Anti-FLT4 Rabbit Polyclonal Antibody
Antibodies
FLT4, also named as VEGFR-3, belongs to the protein kinase superfamily, Tyr protein kinase family and CSF-1/PDGF receptor subfamily. It is a receptor for VEGFC. FLT4 has a tyrosine-protein kinase activity. Defects in FLT4 are the cause of lymphedema hereditary type 1 (LYH1A). Defects in FLT4 are found in juvenile hemangioma. The antibody is specific to FLT4.

Western Blot:PC-3 Cells, 1:200-1:2000

Type: Primary
Antigen: VEGFR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR