Anti-TOP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20705-1-AP-150UL
10096-194EA 402 USD
10096-194
Anti-TOP1 Rabbit Polyclonal Antibody
Antibodies
TOP1 belongs to the eukaryotic type I topoisomerase family. The reaction catalyzed by topoisomerases leads to the conversion of one topological isomer of DNA to another. A chromosomal aberration involving TOP1 is found in a form of therapy-related myelodysplastic syndrome. The antibody is specific to TOP1.

Western Blot:L02 Cells, 1:200-1:2000; IHC: Human cervical cancer, 1:20-1:200

Type: Primary
Antigen: TOP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR