Anti-REPS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20699-1-AP-150UL
10093-516EA 402 USD
10093-516
Anti-REPS2 Rabbit Polyclonal Antibody
Antibodies
REPS2, also named as POB1, is involved in growth factor signaling through its influence on the Ral signaling pathway. The antibody is specific to REPS2.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: REPS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR