Anti-FLNB Rabbit Polyclonal Antibody

Supplier: Proteintech
20685-1-AP-150UL
10087-062EA 402 USD
10087-062
Anti-FLNB Rabbit Polyclonal Antibody
Antibodies
FLNB, also named as FLN1L, FLN3, TABP, TAP and Fh1, belongs to the filamin family. FLNB connects cell membrane constituents to the actin cytoskeleton. It may promote orthogonal branching of actin filaments and links actin filaments to membrane glycoproteins. It is an anchors various transmembrane proteins to the actin cytoskeleton. Interaction with FLNA, FLNB may allow neuroblast migration from the ventricular zone into the cortical plate. Various interactions and localizations of isoforms affect myotube morphology and myogenesis. The antibody is specific to FLNB. It can recognize isoform1,2,3,6,7 of FLNB. It has no cross reaction to FLNA and FLNC.

Western Blot: HeLa Cells, 1:200-1:2000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: FLNB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR