Anti-DOCK3; MOCA Rabbit Polyclonal Antibody

Supplier: Proteintech
20683-1-AP-150UL
10086-062EA 402 USD
10086-062
Anti-DOCK3; MOCA Rabbit Polyclonal Antibody
Antibodies
Dedicator of cytokinesis3 (DOCK3) , also named as MOCA and PBP , is a ~180 kDa protein involved in signaling trasduction. It is a potential guanine nucleotide exchange factor (GEF) which activate some small GTPases by exchanging bound GDP for free GTP. DOCK3 is associated in Alzheimer disease tangles and regulates the accumulation of amyloid precursor protein and beta-amyloid. Overexpression of Dock3 in neural cells promotes axonal outgrowth downstream of brain-derived neurotrophic factor (BDNF) signaling. DOCK3 binds to and inactivates glycogen synthase kinase-3β (GSK-3β) at the plasma membrane, thereby promoteing axon branching and microtubule assembly. By stimulating actin polymerization and microtubule assembly, DOCK3 plays important roles downstream of BDNF signaling in the CNS.

Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: DOCK3; MOCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR