Anti-ABCA2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20681-1-AP-150UL
10082-292EA 402 USD
10082-292
Anti-ABCA2 Rabbit Polyclonal Antibody
Antibodies
ABCA2, also named as ABC2 and KIAA1062, belongs to the ABC transporter ABCA2, a member of the ATP-binding cassette (ABC) transporter family, is primarily expressed in brain and is involved in brain sterol homeostasis and associated with early onset Alzheimer’s disease. ABCA2 is mainly localized in endosomes and lysosomes of oligodendrocytes and Schwann cells. It has also be reported as a marker of neural progenitor in the adult rodent brain. (16539677)

Western Blot: Mouse Thymus Tissue, 1:200-1:2000

Type: Primary
Antigen: ABCA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR