Anti-IL17RB Rabbit Polyclonal Antibody

Supplier: Proteintech
20673-1-AP-150UL
10088-782EA 402 USD
10088-782
Anti-IL17RB Rabbit Polyclonal Antibody
Antibodies
IL17RB, also named as CRL4, EVI27 and IL17BR, is a receptor for the proinflammatory cytokines IL17B and IL17E. IL17RB may play a role in controlling the growth and/or differentiation of hematopoietic cells. IL17RB has two isoforms with MW 56 kDa (isform 1) and 32 kDa (isform 2). The antibody is specific to IL17RB.

Western Blot: Human Testis Tissue, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: IL17RB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR