Anti-NF-M-Specific Rabbit Polyclonal Antibody

Supplier: Proteintech
20664-1-AP-150UL
10090-648EA 402 USD
10090-648
Anti-NF-M-Specific Rabbit Polyclonal Antibody
Antibodies
NEFM, also named as NEF3 and NFM, belongs to the intermediate filament family. Neurofilaments are the 10nm intermediate filaments found specifically in neurons. They are a major component of the cell's cytoskeleton, and provide support for normal axonal radial growth. Neurofilaments usually contain three intermediate filament proteins: L, M, and H which are involved in the maintenance of neuronal caliber. The names given to the three major neurofilament subunits are based upon the apparent molecular weight of the mammalian subunits on SDS-PAGE:NF-L, 65-68kd; NF-M,145-160kd and NF-H, 200-220kd.The antibody is specific to NEFM.

IF: Hela Cells, 1:10-1:100; Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC:rat Brain Tissue, 1:20-1:200

Type: Primary
Antigen: NF-M-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR