Anti-MARCKS Rabbit Polyclonal Antibody

Supplier: Proteintech
20661-1-AP-150UL
10089-980EA 402 USD
10089-980
Anti-MARCKS Rabbit Polyclonal Antibody
Antibodies
MARCKS, also named as MACS, PRKCSL and 80K-L protein, belongs to the MARCKS family. MARCKS is the most prominent cellular substrate for protein kinase C. It binds calmodulin, actin, and synapsin. MARCKS is a filamentous (F) actin cross-linking protein.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human Heart, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: MARCKS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR