Anti-NPTX1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20656-1-AP-150UL
10090-908EA 402 USD
10090-908
Anti-NPTX1 Rabbit Polyclonal Antibody
Antibodies
NPTX1, also named as NP1, may mediate uptake of degraded synaptic material which could play an important role in synaptic remodeling. NPTX1 can mediate the neuronal and glial uptake of the snake venom toxin taipoxin. The antibody is specific to NPTX1.

Western Blot: HepG2 Cells, 1:200-1:2000; IF: Hela, 1:10-1:100

Type: Primary
Antigen: NPTX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR