Anti-ANO2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20647-1-AP-150UL
10082-940EA 402 USD
10082-940
Anti-ANO2 Rabbit Polyclonal Antibody
Antibodies
ANO2, also named as C12orf3 and TMEM16B, belongs to the anoctamin family. It acts as a calcium-activated chloride channel (CaCC), mostly in photoreceptors. It may mediate olfactory amplification in olfactory sensory neurons (OSNs) and light perception amplification in retina.

Western Blot: Mouse Brain Tissue, 1:1000-1:10000; IF: HepG2 Cells, 1:10-1:100; IP: Mouse Brain Tissue, 1:200-1:1000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ANO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR