Anti-PPAPDC3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20635-1-AP-150UL
10092-624EA 402 USD
10092-624
Anti-PPAPDC3 Rabbit Polyclonal Antibody
Antibodies
PPAPDC3(Phosphatidic acid phosphatase type 2 domain-containing protein 3) is also named as C9orf67 and belongs to the PA-phosphatase related phosphoesterase family. It plays a role as negative regulator of myoblast differentiation, in part through effects on MTOR signaling.

Western Blot: Mouse Skeletal Muscle Tissue, 1:200-1:2000

Type: Primary
Antigen: PPAPDC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR