Anti-CXCR2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20634-1-AP-150UL
10085-428EA 402 USD
10085-428
Anti-CXCR2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:L02 Cells, 1:1000-1:10000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CXCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR