Anti-ZIP4 Rabbit Polyclonal Antibody

Supplier: Proteintech
20625-1-AP-150UL
10097-290EA 537.7 USD
10097-290
Anti-ZIP4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: ZIP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR