Anti-DIAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20624-1-AP-150UL
10085-852EA 402 USD
10085-852
Anti-DIAP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:1000-1:10000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: DIAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR