Anti-ABHD12B Rabbit Polyclonal Antibody

Supplier: Proteintech
20623-1-AP-150UL
10082-358EA 402 USD
10082-358
Anti-ABHD12B Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse skin Tissue, 1:200-1:2000; IHC: Human skin, 1:20-1:200

Type: Primary
Antigen: ABHD12B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR