Anti-PEX7 Rabbit Polyclonal Antibody

Supplier: Proteintech
20614-1-AP-150UL
10091-466EA 402 USD
10091-466
Anti-PEX7 Rabbit Polyclonal Antibody
Antibodies
PEX7, also known as peroxisomal targeting signal 2 (PTS2) receptor, is one member of peroxins (PEXs) which are proteins required for peroxisome assembly. PEX5 and PEX7 function as receptors that recognize PTS1- and PTS2- containing proteins, respectively. PEX7 plays an essential role in peroxisomal protein import by binding to the N-terminal PTS2-type peroxisomal targeting signal. PEX7 defects are associated with peroxisome biogenesis disorder complementation group 11 (PBD-CG11), rhizomelic chondrodysplasia punctata type 1 (RCDP1) and Refsum disease.

Western Blot: MDA-MB-453s Cells, 1:500-1:5000

Type: Primary
Antigen: PEX7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR