Anti-HECTD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20605-1-AP-150UL
10088-146EA 402 USD
10088-146
Anti-HECTD1 Rabbit Polyclonal Antibody
Antibodies
HECTD1 (E3 ubiquitin-protein ligase), which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates,may be required for development of the head mesenchyme and neural tube closure.It is also involved in sporadic Parkinson’s disease.This protein is specific to HECTD1.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: HECTD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR