Anti-ORAI2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20592-1-AP-150UL
10091-598EA 402 USD
10091-598
Anti-ORAI2 Rabbit Polyclonal Antibody
Antibodies
ORAI2, also named as C7orf19, CBCIP2, TMEM142B and PP1729, is a Ca2+ release-activated Ca2+-like (CRAC-like) channel subunit. The MW of ORAI2 is about 36 kDa for glycosylation .

Western Blot: MCF-7 Cells, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200; IF: HEK-293 Cells, 1:10-1:100; FC: HEK-293 Cells, N/A

Type: Primary
Antigen: ORAI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR