Anti-CHD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20576-1-AP-150UL
10084-434EA 402 USD
10084-434
Anti-CHD1 Rabbit Polyclonal Antibody
Antibodies
CHD1 belongs to the SNF2/RAD54 helicase family. CHD1 is an aTP-dependent chromatin-remodeling factor which functions as substrate recognition component of the transcription regulatory histone acetylation (HAT) complex SAGA. CHD1 regulates polymerase II transcription. It is required for efficient transcription by RNA polymerase I, and more specifically the polymerase I transcription termination step. CHD1 regulates negatively DNA replication. It is required to maintain a specific chromatin configuration across the genome. It is associated with histone deacetylase (HDAC) activity. CHD1 is required for the bridging of SNF2, the FACT complex, the PAF complex as well as the U2 snRNP complex to H3K4me3. It functions to modulate the efficiency of pre-mRNA splicing in part through physical bridging of spliceosomal components to H3K4me3. It is required for maintaining open chromatin and pluripotency in embryonic stem cells. The antibody is specific to CHD1.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human breast cancer, 1:20-1:200

Type: Primary
Antigen: CHD1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR