Anti-DOC2B Rabbit Polyclonal Antibody

Supplier: Proteintech
20574-1-AP-150UL
10086-056EA 402 USD
10086-056
Anti-DOC2B Rabbit Polyclonal Antibody
Antibodies
DOC2B may be involved in constitutive membrane trafficking including dynein-dependent intracellular vesicle transport. In vitro, binds calcium and phospholipids. The antibody is specific to DOC2B.

Western Blot: Mouse Kidney Tissue, 1:200-1:2000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: DOC2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR