Anti-RAC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20573-1-AP-150UL
10093-268EA 402 USD
10093-268
Anti-RAC2 Rabbit Polyclonal Antibody
Antibodies
RAC2, also named as GX, belongs to the small GTPase superfamily and Rho family. RAC2 is a plasma membrane-associated small GTPase which cycles between an active GTP-bound and inactive GDP-bound state. In active state binds to a variety of effector proteins to regulate cellular responses, such as secretory processes, phagocytose of apoptotic cells and epithelial cell polarization. RAC2 augments the production of reactive oxygen species (ROS) by NADPH oxidase. Defects in RAC2 are the cause of neutrophil immunodeficiency syndrome. The antibody is specific to RAC2.

Western Blot:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: RAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR