Anti-PRDX2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20569-1-AP-150UL
10091-872EA 402 USD
10091-872
Anti-PRDX2 Rabbit Polyclonal Antibody
Antibodies
PRDX2, also named as NKEFB, TDPX1, TSA and PRP, belongs to the ahpC/TSA family. It is involved in redox regulation of the cell. Reduces peroxides with reducing equivalents provided through the thioredoxin system. It is not able to receive electrons from glutaredoxin. PRDX2 may play an important role in eliminating peroxides generated during metabolism. It might participate in the signaling cascades of growth factors and tumor necrosis factor-alpha by regulating the intracellular concentrations of H2O2. The antibody is specific to PRDX2.

Western Blot: HeLa Cells, 1:500-1:5000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: PRDX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR