Anti-PAFAH1B3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20564-1-AP-150UL
10091-740EA 402 USD
10091-740
Anti-PAFAH1B3 Rabbit Polyclonal Antibody
Antibodies
PAFAH1B3, also named as PAFAHG, belongs to the 'GDSL' lipolytic enzyme family and Platelet-activating factor acetylhydrolase IB beta/gamma subunits subfamily. It inactivates paf by removing the acetyl group at the sn-2 position. This is a catalytic subunit. PAFAH1B3 plays an important role during the development of brain. The antibody is specific to PAFAH1B3.

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human spleen Tissue, 1:20-1:200

Type: Primary
Antigen: PAFAH1B3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR