Anti-STXBP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20562-1-AP-150UL
10081-134EA 402 USD
10081-134
Anti-STXBP1 Rabbit Polyclonal Antibody
Antibodies
STXBP1, also named as UNC18A, N-Sec1 and p67 belongs to the STXBP/unc-18/SEC1 family. STXBP1 may participate in the regulation of synaptic vesicle docking and fusion, possibly through interaction with GTP-binding proteins. It is essential for neurotransmission and binds syntaxin, a component of the synaptic vesicle fusion machinery probably in a 1:1 ratio. STXBP1 can interact with syntaxins 1, 2, and 3 but not syntaxin 4. It may play a role in determining the specificity of intracellular fusion reactions. The antibody is specific to STXBP1.

Western Blot: Human Brain Tissue, 1:1000-1:10000; IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: STXBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR