Anti-CBLN4 Rabbit Polyclonal Antibody

Supplier: Proteintech
20559-1-AP-150UL
10084-130EA 402 USD
10084-130
Anti-CBLN4 Rabbit Polyclonal Antibody
Antibodies
CBLN4 may be involved in synaptic functions in the CNS. CBLN4 can enable ER export and secretion of CBLN3. The antibody has no cross reaction to CBLN1, 2, or 3.

Western Blot: Mouse cerebellum Tissue, 1:200-1:1000; IHC: Human cerebellum Tissue, 1:20-1:200

Type: Primary
Antigen: CBLN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR