Anti-TOP2B Rabbit Polyclonal Antibody

Supplier: Proteintech
20549-1-AP-150UL
10096-200EA 402 USD
10096-200
Anti-TOP2B Rabbit Polyclonal Antibody
Antibodies
TOP2B belongs to the type II topoisomerase family. TOP2B control topological states of DNA by transient breakage and subsequent rejoining of DNA strands. TOP2B makes double-strand breaks. It is indirectly ivolved in vitamin D-coupled transcription regulation via its association with the WINAC complex, a chromatin-remodeling complex recruited by vitamin D receptor (VDR), which is required for the ligand-bound VDR-mediated transrepression of the CYP27B1 gene. The antibody is specific to TOP2B.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human cervical cancer, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: TOP2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR