Anti-ERCC6 Rabbit Polyclonal Antibody

Supplier: Proteintech
20548-1-AP-150UL
10081-298EA 402 USD
10081-298
Anti-ERCC6 Rabbit Polyclonal Antibody
Antibodies
ERCC6, also named as CSB, belongs to the SNF2/RAD54 helicase family. It is involved in the preferential repair of active genes. It is presumed DNA or RNA unwinding function. ERCC6 corrects the UV survival and RNA synthesis after UV exposure of Cockayne syndrome complementation group B. Defects in ERCC6 are the cause of Cockayne syndrome type B (CSB) , cerebro-oculo-facio-skeletal syndrome type 1 (COFS1), De Sanctis-Cacchione syndrome (DSC), and UV-sensitive syndrome (UVS). Genetic variation in ERCC6 is associated with susceptibility to age-related macular degeneration type 5 (ARMD5). The antibody is specific to ERCC6.

Western Blot: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: ERCC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR