Anti-NUDT3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20542-1-AP-150UL
10091-380EA 402 USD
10091-380
Anti-NUDT3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse cerebellum Tissue, 1:200-1:2000

Type: Primary
Antigen: NUDT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR