Anti-SH3BP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20541-1-AP-150UL
10094-530EA 402 USD
10094-530
Anti-SH3BP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse lung Tissue, 1:1000-1:10000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: SH3BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR