Anti-EFS Rabbit Polyclonal Antibody

Supplier: Proteintech
20538-1-AP-150UL
10086-300EA 402 USD
10086-300
Anti-EFS Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Thymus Tissue, 1:500-1:5000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: EFS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR