Anti-LAYN Rabbit Polyclonal Antibody

Supplier: Proteintech
20535-1-AP-150UL
10089-540EA 402 USD
10089-540
Anti-LAYN Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: LAYN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR