Anti-ZXDC Rabbit Polyclonal Antibody

Supplier: Proteintech
20530-1-AP-150UL
10097-410EA 402 USD
10097-410
Anti-ZXDC Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Kidney Tissue, 1:200-1:2000; IF: Hela, 1:10-1:100

Type: Primary
Antigen: ZXDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR