Anti-IL20RB Rabbit Polyclonal Antibody

Supplier: Proteintech
20521-1-AP-150UL
10088-810EA 402 USD
10088-810
Anti-IL20RB Rabbit Polyclonal Antibody
Antibodies
Interleukin-20 receptor subunit beta (IL20RB) is a single-pass type I membrane protein of the type II cytokine receptor family. The interleukin-20-receptor I complex (IL-20-RI) is composed of two chains, IL20RA and IL20RB. IL-20-RI is a receptor for IL-19, IL-20 and IL-24. IL20RB can also form a heterodimer with IL22RA1. The IL22RA1/IL20RB dimer is a receptor for IL20 and IL24.

Western Blot: MCF7 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: IL20RB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR