Anti-CEPT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20496-1-AP-150UL
10084-666EA 402 USD
10084-666
Anti-CEPT1 Rabbit Polyclonal Antibody
Antibodies
IHC: Human pancreas, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: CEPT1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR