Anti-TMEM165 Rabbit Polyclonal Antibody

Supplier: Proteintech
20485-1-AP-150UL
10096-088EA 402 USD
10096-088
Anti-TMEM165 Rabbit Polyclonal Antibody
Antibodies
Thiopurine methyltransferase or thiopurine S-methyltransferase (TPMT), a member of methyltransferase superfamily, is an enzyme involved in the normal metabolic inactivation of thiopurine drugs such as azathioprine, 6-mercaptopurine and 6-thioguanine. These drugs are generally used as immunosupressants or cytotoxic drugs and are prescribed for a variety of clinical conditions including leukemia, autoimmune disease and organ transplantation. Defects in the TPMT gene leads to decreased methylation and decreased inactivation of 6MP leading to enhanced bone marrow toxicity which may cause myelosuppression, anemia, bleeding tendency, leukopenia and infection.

Western Blot:Jurkat Cells, 1:1000-1:10000; IF: HepG2 Cells, 1:20-1:200; IP: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: TMEM165
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR