Anti-TBC1D17 Rabbit Polyclonal Antibody

Supplier: Proteintech
20482-1-AP-150UL
10095-624EA 537.7 USD
10095-624
Anti-TBC1D17 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:500-1:5000; IF: MCF-7 Cells, 1:20-1:200; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TBC1D17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR