Anti-ZNF34 Rabbit Polyclonal Antibody

Supplier: Proteintech
20477-1-AP-150UL
10097-336EA 402 USD
10097-336
Anti-ZNF34 Rabbit Polyclonal Antibody
Antibodies
ZNF34, also named as KOX32, contains twelve C2H2-type zinc fingers and one KRAB domain. It belongs to the krueppel C2H2-type zinc-finger protein family. This protein may be involved in transcriptional regulation.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: ZNF34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR