Anti-C9orf156 Rabbit Polyclonal Antibody

Supplier: Proteintech
20475-1-AP-150UL
10083-896EA 402 USD
10083-896
Anti-C9orf156 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: C9orf156
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR