Anti-CK34 Rabbit Polyclonal Antibody

Supplier: Proteintech
20449-1-AP-150UL
10089-392EA 402 USD
10089-392
Anti-CK34 Rabbit Polyclonal Antibody
Antibodies
Keratins are a large family of proteins that form the intermediate filament cytoskeleton of epithelial cells. Keratin expression is highly regulated, tissue specific, and varies according to cell-state. Type I keratins consist of acidic, low molecular weight proteins with MW ranging from 40 kDa (KRT19) to 64 kDa (KRT9). Type 2 keratins consist of basic or neutral, high molecular weight proteins with MW from 52 kDa (KRT8) to 67 kDa (KRT18). KRT 34 is a type I keratin.

Western Blot:A431 Cells, 1:1000-1:10000; IHC: Human skin, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: CK34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR