Anti-USP33 Rabbit Polyclonal Antibody

Supplier: Proteintech
20445-1-AP-150UL
10081-328EA 402 USD
10081-328
Anti-USP33 Rabbit Polyclonal Antibody
Antibodies
USP33(Ubiquitin carboxyl-terminal hydrolase 33) is also named as KIAA1097, VDU1 and belongs to the peptidase C19 family. This protein, originally identified as a von Hippel-Lindau tumor suppressor (VHL) protein-interacting deubiquitinating enzyme, binds to the Robo1 receptor, and that USP33 is required for Slit responsiveness in breast cancer cells. The localization of USP33 is broadly confined to the secretory pathway, with all variants localizing to endoplasmic reticulum-associated structures. It has 3 isoforms produced by alternative splicing. This antibody is specific to USP33.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human Prostate Cancer Tissue, 1:20-1:200

Type: Primary
Antigen: USP33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR